Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smg5
Entrez Gene ID
229512
Gene Name
Smg-5 homolog, nonsense mediated mRNA decay factor (C. elegans)
Gene Aliases
BC024683, mKIAA1089
Chromosome Location
Chr.3: 88336260 - 88362338 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178246.3 NP_839977.2 19 - 20 2923 68
GenBank mRNA AK129286.1 - 19 - 20 2902 68
AK133688.1 - 10 - 11 1747 68
AK143911.1 - 19 - 20 2925 68
AK172217.1 - 18 - 19 2803 68
BC024089.1 - 2 - 3 212 68
BC048378.1 - 8 - 9 1103 68

View Full Product Details