Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhdc1
Entrez Gene ID
271005
Gene Name
kelch domain containing 1
Gene Aliases
-
Chromosome Location
Chr.12: 69226917 - 69284646 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178253.5 NP_839984.1 5 - 6 572 94
GenBank mRNA AK144621.1 - 5 - 6 512 94
AY238938.1 - 5 - 6 572 94
BC119567.2 - 5 - 6 556 88
BC119568.2 - 5 - 6 556 94

View Full Product Details