The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178756.4 | NP_848871.1 | 3 - 4 | 451 | 93 |
GenBank mRNA | AK034251.1 | - | 3 - 4 | 636 | 93 |
AK043762.1 | - | 3 - 4 | 353 | 93 | |
AK047614.1 | - | 3 - 4 | 381 | 93 | |
AK053451.1 | - | 3 - 4 | 457 | 93 | |
AK087439.1 | - | 3 - 4 | 602 | 93 | |
AK087514.1 | - | 3 - 4 | 452 | 93 | |
AK160897.1 | - | 3 - 4 | 578 | 93 | |
AY345342.1 | - | 3 - 4 | 451 | 93 | |
BC096762.1 | - | 3 - 4 | 427 | 93 |