The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252458.1 | NP_001239387.1 | 2 - 3 | 521 | 70 |
NM_178900.4 | NP_849231.1 | 1 - 2 | 806 | 70 | |
GenBank mRNA | AK037030.1 | - | 1 - 2 | 807 | 70 |
AK154029.1 | - | 1 - 2 | 655 | 70 | |
AK170236.1 | - | 2 - 3 | 494 | 70 | |
AK170761.1 | - | 2 - 3 | 485 | 70 | |
BC095949.1 | - | 2 - 3 | 471 | 70 | |
BC096444.1 | - | 2 - 3 | 489 | 70 |