The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081240.3 | NP_001074709.1 | 2 - 3 | 492 | 88 |
GenBank mRNA | AK045296.1 | - | 2 - 3 | 483 | 88 |
AK045315.1 | - | 2 - 3 | 482 | 88 | |
AK154546.1 | - | 2 - 3 | 436 | 88 | |
AK161023.1 | - | 2 - 3 | 436 | 88 |