Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbl2
Entrez Gene ID
27368
Gene Name
transducin (beta)-like 2
Gene Aliases
C76179, WS-bTRP
Chromosome Location
Chr.5: 135149426 - 135163876 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013763.2 NP_038791.2 3 - 4 459 70
GenBank mRNA AK132366.1 - 3 - 4 485 70
AK165686.1 - 3 - 4 479 70
BC039273.1 - 3 - 4 492 70
BC048185.1 - 3 - 4 459 70

View Full Product Details