Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Apln
Entrez Gene ID
30878
Gene Name
apelin
Gene Aliases
6030430G11Rik, Apel
Chromosome Location
Chr.X: 48025146 - 48034852 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013912.3 NP_038940.1 2 - 3 583 71
GenBank mRNA AK042076.1 - 2 - 3 587 71
BC020015.1 - 2 - 3 564 71
DQ832282.1 - 2 - 3 572 71

View Full Product Details