Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpsf2
Entrez Gene ID
51786
Gene Name
cleavage and polyadenylation specific factor 2
Gene Aliases
100kDa, 2610024B04Rik, AI662483, Cpsf, MCPSF, mKIAA1367
Chromosome Location
Chr.12: 101975765 - 102006430 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016856.3 NP_058552.1 9 - 10 1440 120
GenBank mRNA AF012822.1 - 9 - 10 1426 120
AK140869.1 - 9 - 10 1422 120
BC007163.1 - 7 - 8 1098 120
BC013628.1 - 9 - 10 1415 120

View Full Product Details