Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hr
Entrez Gene ID
15460
Gene Name
hairless
Gene Aliases
ALUNC, AU, N, ba, bldy, hr, rh, rh-bmh, rhino
Chromosome Location
Chr.14: 70554056 - 70573548 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021877.3 NP_068677.2 1 - 2 57 55
GenBank mRNA BC049182.1 - 1 - 2 57 55

View Full Product Details