The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053014.3 | NP_443747.2 | 2 - 3 | 140 | 55 |
GenBank mRNA | AK008965.1 | - | 2 - 3 | 298 | 55 |
AK075715.1 | - | 2 - 3 | 140 | 55 | |
AK196461.1 | - | 2 - 3 | 89 | 55 |