Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map7d1
Entrez Gene ID
245877
Gene Name
MAP7 domain containing 1
Gene Aliases
AV028413, BC019977, Mtap7d1, Parcc1, Rprc1, mKIAA1187
Chromosome Location
Chr.4: 126232167 - 126256417 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145970.1 NP_001139442.1 15 - 16 2381 139
NM_144941.3 NP_659190.3 15 - 16 2597 139
GenBank mRNA AK122462.1 - 7 - 8 4303 139
AK151613.1 - 13 - 14 2112 139
BC016081.1 - 6 - 7 633 139
BC019977.1 - 10 - 11 1573 139
BC023677.1 - 15 - 16 2572 139
BC023774.1 - 15 - 16 2378 139
BM944422.1 - 6 - 7 487 139

View Full Product Details