Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep68
Entrez Gene ID
216543
Gene Name
centrosomal protein 68
Gene Aliases
6030463E10Rik, AI481761, BC027174, Kiaa0582
Chromosome Location
Chr.11: 20226688 - 20250127 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172260.3 NP_758464.2 1 - 2 117 91
GenBank mRNA AK031622.1 - 1 - 2 114 91
AK140820.1 - 1 - 2 118 91
AK146669.1 - 1 - 2 65 91
AK162594.1 - 1 - 2 123 91
AK169934.1 - 1 - 2 113 91
AK172304.1 - 1 - 2 78 91
BC027174.1 - 1 - 2 111 91

View Full Product Details