Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep68
Entrez Gene ID
216543
Gene Name
centrosomal protein 68
Gene Aliases
6030463E10Rik, AI481761, BC027174, Kiaa0582
Chromosome Location
Chr.11: 20226688 - 20250127 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172260.3 NP_758464.2 3 - 4 1968 67
GenBank mRNA AK031622.1 - 3 - 4 1965 67
AK140820.1 - 3 - 4 1969 67
AK148267.1 - 1 - 2 1108 67
AK169934.1 - 3 - 4 1966 67
BC027174.1 - 3 - 4 1962 67

View Full Product Details