Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbce
Entrez Gene ID
70430
Gene Name
tubulin-specific chaperone E
Gene Aliases
2610206D02Rik, C530005D02Rik, pmn
Chromosome Location
Chr.13: 13997949 - 14039638 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK011899.1 - 2 - 3 213 57
AK082940.1 - 2 - 3 233 57
AK167383.1 - 2 - 3 221 57
AK167848.1 - 2 - 3 217 57
AY082332.1 - 2 - 3 187 57
BC050206.1 - 2 - 3 195 57

View Full Product Details