The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011119.3 | NP_035249.1 | 7 - 8 | 739 | 102 |
GenBank mRNA | AK076286.1 | - | 7 - 8 | 744 | 102 |
AK144645.1 | - | 7 - 8 | 793 | 102 | |
AK166936.1 | - | 7 - 8 | 748 | 102 | |
AK168582.1 | - | 7 - 8 | 741 | 102 | |
AK207779.1 | - | 3 - 4 | 198 | 102 | |
BC003777.1 | - | 7 - 8 | 729 | 102 | |
BC005593.1 | - | 7 - 8 | 729 | 102 | |
BC028926.1 | - | 7 - 8 | 722 | 102 | |
BC037623.1 | - | 7 - 8 | 759 | 102 | |
BC046532.1 | - | 7 - 8 | 693 | 102 | |
BC086788.1 | - | 5 - 6 | 2945 | 102 | |
U43918.1 | - | 7 - 8 | 898 | 102 | |
X84789.1 | - | 7 - 8 | 804 | 102 |