Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr13
Entrez Gene ID
73447
Gene Name
WD repeat domain 13
Gene Aliases
1700060B08Rik, 5730411P10Rik, DXHXS7467e, W51679, mMg21
Chromosome Location
Chr.X: 8123301 - 8132858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290783.1 NP_001277712.1 3 - 4 888 62
NM_026137.5 NP_080413.2 3 - 4 922 62
NM_028542.1 NP_082818.1 3 - 4 1174 62
GenBank mRNA AF220146.1 - 2 - 3 444 62
AF353243.1 - 3 - 4 568 62
AF513713.1 - 3 - 4 391 62
AK006837.1 - 2 - 3 410 62
AK017549.1 - 3 - 4 504 62
BC037502.1 - 3 - 4 539 62
BC057366.1 - 3 - 4 888 62
BC080853.1 - 3 - 4 837 62
BC108332.1 - 3 - 4 678 62
BC120553.1 - 3 - 4 425 62
BC125287.1 - 3 - 4 424 62

View Full Product Details