The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007387.2 | NP_031413.1 | 10 - 11 | 1159 | 95 |
GenBank mRNA | AK089401.1 | - | 10 - 11 | 1150 | 95 |
AK128963.1 | - | 10 - 11 | 1152 | 95 | |
AK170658.1 | - | 10 - 11 | 1160 | 95 | |
BC013495.1 | - | 10 - 11 | 1147 | 95 | |
BC023343.1 | - | 10 - 11 | 1141 | 95 | |
X57199.1 | - | 10 - 11 | 1130 | 95 |