Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nudt11
Entrez Gene ID
58242
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 11
Gene Aliases
DIPP3, DIPP3b
Chromosome Location
Chr.X: 6047479 - 6054751 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021431.2 NP_067406.2 1 - 2 895 116
GenBank mRNA AB041576.1 - 1 - 2 641 116
AK036077.1 - 1 - 2 226 116
AK053310.1 - 1 - 2 675 116
AY152853.1 - 1 - 2 641 116
BC071274.1 - 1 - 2 699 116

View Full Product Details