The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025288.2 | NP_079564.1 | 2 - 3 | 204 | 138 |
GenBank mRNA | AK014349.1 | - | 2 - 3 | 207 | 138 |
AY169236.1 | - | 2 - 3 | 187 | 138 | |
BC107224.1 | - | 2 - 3 | 198 | 138 | |
BC107225.1 | - | 2 - 3 | 200 | 138 | |
M92419.1 | - | 2 - 3 | 188 | 138 |