The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025450.4 | NP_079726.1 | 3 - 4 | 411 | 112 |
GenBank mRNA | AB049951.1 | - | 2 - 3 | 370 | 112 |
AK003008.1 | - | 3 - 4 | 228 | 112 | |
AK007356.1 | - | 3 - 4 | 412 | 112 | |
AK009568.1 | - | 3 - 4 | 409 | 112 | |
AK134065.1 | - | 2 - 3 | 1197 | 112 | |
BC003890.1 | - | 2 - 3 | 348 | 112 |