The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130857.2 | NP_570927.1 | 1 - 1 | 387 | 86 |
GenBank mRNA | AF345295.1 | - | 1 - 1 | 339 | 86 |
AK020697.1 | - | 1 - 1 | 388 | 86 | |
BC115545.2 | - | 1 - 1 | 339 | 86 | |
BC115546.2 | - | 1 - 1 | 339 | 86 |