The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133209.2 | NP_573472.2 | 4 - 5 | 799 | 147 |
GenBank mRNA | AJ400847.1 | - | 4 - 5 | 748 | 147 |
BC145880.1 | - | 4 - 5 | 737 | 147 | |
BC145906.1 | - | 4 - 5 | 737 | 147 |