The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009487.1 | NP_033513.1 | 3 - 4 | 1378 | 97 |
GenBank mRNA | AF011421.1 | - | 3 - 4 | 1378 | 97 |
BC145855.1 | - | 3 - 4 | 1339 | 97 |