The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010731.3 | NP_034861.3 | 2 - 3 | 1686 | 144 |
GenBank mRNA | AF086830.1 | - | 2 - 3 | 1303 | 144 |
AK010379.1 | - | 1 - 2 | 174 | 144 | |
AK149961.1 | - | 2 - 3 | 1364 | 144 | |
AK154907.1 | - | 2 - 3 | 1380 | 144 | |
BC024815.2 | - | 2 - 3 | 1686 | 144 | |
BC057204.1 | - | 2 - 3 | 1324 | 144 | |
BC138524.1 | - | 2 - 3 | 1445 | 144 | |
BC145311.1 | - | 2 - 3 | 1485 | 144 | |
BC171957.1 | - | 2 - 3 | 1445 | 144 |