Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc66
Entrez Gene ID
320234
Gene Name
coiled-coil domain containing 66
Gene Aliases
E230015L20Rik
Chromosome Location
Chr.14: 27481090 - 27508506 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177111.3 NP_796085.3 17 - 18 2749 137
GenBank mRNA AK053185.1 - 2 - 3 299 137
AK087583.1 - 9 - 10 1470 137
AK158666.1 - 15 - 16 2267 137
AK170006.1 - 15 - 16 2216 137
BC070471.1 - 17 - 18 2750 137
BC072583.1 - 9 - 10 1866 137

View Full Product Details