The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019916.2 | NP_064300.2 | 1 - 2 | 554 | 163 |
GenBank mRNA | AJ223801.1 | - | 1 - 2 | 425 | 163 |
AK141870.1 | - | 1 - 2 | 555 | 163 | |
BC141308.1 | - | 1 - 2 | 484 | 163 | |
BC145631.1 | - | 1 - 2 | 484 | 163 |