Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Endov
Entrez Gene ID
338371
Gene Name
endonuclease V
Gene Aliases
A730011L01Rik
Chromosome Location
Chr.11: 119491323 - 119511465 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164636.1 NP_001158108.1 6 - 7 643 58
NM_177394.3 NP_796368.1 5 - 6 695 58
GenBank mRNA AK042626.1 - 5 - 6 583 58
AK084233.1 - 6 - 7 645 58
AK084548.1 - 6 - 7 519 58
AK170595.1 - 5 - 6 688 58
AK172181.1 - 5 - 6 662 58
BC056486.1 - 5 - 6 615 58

View Full Product Details