Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grpel2
Entrez Gene ID
17714
Gene Name
GrpE-like 2, mitochondrial
Gene Aliases
mt-GrpE#2, mt-Grpel2
Chromosome Location
Chr.18: 61712424 - 61726361 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK082983.1 - - 2066 77

View Full Product Details