Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ndc80
Entrez Gene ID
67052
Gene Name
NDC80 homolog, kinetochore complex component (S. cerevisiae)
Gene Aliases
2610020P18Rik, AV139730, HEC, HEC1, Kntc2
Chromosome Location
Chr.17: 71496100 - 71526857 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023294.2 NP_075783.2 14 - 15 1707 123
GenBank mRNA AF326730.1 - 14 - 15 1720 123
AK011497.1 - 14 - 15 1677 123
AK011554.1 - 10 - 11 1103 123
AK163317.1 - 14 - 15 1658 123
AK169427.1 - 14 - 15 1664 123
BC020131.1 - 14 - 15 1643 123

View Full Product Details