The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027453.2 | NP_081729.1 | 5 - 6 | 519 | 130 |
GenBank mRNA | AK004239.1 | - | 5 - 6 | 550 | 130 |
AK011367.1 | - | 5 - 6 | 865 | 130 | |
AK013120.1 | - | 5 - 6 | 507 | 130 | |
AK076283.1 | - | 5 - 6 | 519 | 130 | |
BC024612.1 | - | 5 - 6 | 502 | 130 | |
BC024920.1 | - | 5 - 6 | 462 | 130 | |
BC058282.1 | - | 5 - 6 | 475 | 130 | |
BC099407.1 | - | 4 - 5 | 454 | 130 | |
BC138215.1 | - | 5 - 6 | 500 | 130 | |
BC138216.1 | - | 5 - 6 | 500 | 130 |