Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dsg3
Entrez Gene ID
13512
Gene Name
desmoglein 3
Gene Aliases
bal, balding
Chromosome Location
Chr.18: 20510304 - 20541310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030596.3 NP_085099.2 4 - 5 438 70
GenBank mRNA AK029018.1 - - 460 70
AK037142.1 - 4 - 5 469 70
AK148657.1 - 4 - 5 439 70
BC132058.1 - 4 - 5 467 70
BC132639.1 - 4 - 5 467 70
U86016.1 - 4 - 5 367 70

View Full Product Details