The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133357.3 | NP_579935.1 | 2 - 3 | 787 | 158 |
GenBank mRNA | AF343088.1 | - | 1 - 2 | 205 | 158 |
AK028972.1 | - | 2 - 3 | 788 | 158 | |
AK037233.1 | - | 2 - 3 | 788 | 158 | |
AK076367.1 | - | 2 - 3 | 788 | 158 | |
BC137935.1 | - | 2 - 3 | 788 | 158 | |
BC137936.1 | - | 2 - 3 | 788 | 158 |