Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Commd7
Entrez Gene ID
99311
Gene Name
COMM domain containing 7
Gene Aliases
2310010I22Rik, AW060956, mU3
Chromosome Location
Chr.2: 153616930 - 153632794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195390.1 NP_001182319.1 4 - 5 421 95
NM_133850.2 NP_598611.2 3 - 4 346 95
GenBank mRNA AK008447.1 - 3 - 4 301 95
AK009274.1 - 3 - 4 321 95
AK046582.1 - 3 - 4 296 95
AK077349.1 - 3 - 4 287 95
AK078774.1 - 3 - 4 305 95
AK164805.1 - 3 - 4 255 95
AK169037.1 - 4 - 5 368 95
BC003310.1 - 3 - 4 279 95
BC065794.1 - 3 - 4 247 95
BC092215.1 - 3 - 4 263 95
BC094216.1 - 3 - 4 303 95
BY295213.1 - 3 - 4 347 95

View Full Product Details