The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133913.2 | NP_598674.2 | 1 - 2 | 1550 | 110 |
GenBank mRNA | AK089404.1 | - | 1 - 2 | 1560 | 110 |
AK122504.1 | - | 1 - 2 | 1550 | 110 | |
AK138814.1 | - | - | 1556 | 110 | |
BC047275.1 | - | 1 - 2 | 773 | 110 |