Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lsr
Entrez Gene ID
54135
Gene Name
lipolysis stimulated lipoprotein receptor
Gene Aliases
ILDR3, Lisch7
Chromosome Location
Chr.7: 30957770 - 30973469 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164184.1 NP_001157656.1 2 - 3 740 57
NM_001164185.1 NP_001157657.1 2 - 3 740 57
NM_017405.2 NP_059101.1 2 - 3 740 57
GenBank mRNA AK146807.1 - 2 - 3 456 57
AK167463.1 - 2 - 3 679 57
BC004672.1 - 2 - 3 486 57
U49507.1 - 2 - 3 631 57

View Full Product Details