Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rsl1d1
Entrez Gene ID
66409
Gene Name
ribosomal L1 domain containing 1
Gene Aliases
2410005K20Rik, C76439, pBK1
Chromosome Location
Chr.16: 11193037 - 11203292 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025546.2 NP_079822.1 4 - 5 570 114
GenBank mRNA AK012937.1 - 4 - 5 563 114
AK075933.1 - 4 - 5 571 114
AK136025.1 - 4 - 5 537 114
AK145699.1 - 4 - 5 584 114
AK162241.1 - 4 - 5 544 114
AK165572.1 - 4 - 5 583 114
AK166479.1 - 4 - 5 114
AK166579.1 - 4 - 5 566 114
AK168469.1 - 4 - 5 566 114
AK168717.1 - 4 - 5 559 114
AK171805.1 - 4 - 5 558 114
BC034355.1 - 4 - 5 536 114
BC069934.1 - 4 - 5 544 114
DQ445383.1 - 4 - 5 578 114

View Full Product Details