The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025648.3 | NP_079924.2 | 2 - 3 | 332 | 89 |
GenBank mRNA | AK002577.1 | - | 2 - 3 | 309 | 89 |
AK031697.1 | - | 2 - 3 | 310 | 89 | |
AK044637.1 | - | 2 - 3 | 310 | 89 | |
AK084031.1 | - | 2 - 3 | 294 | 89 | |
AK154629.1 | - | 2 - 3 | 330 | 89 | |
BB853717.1 | - | 2 - 3 | 334 | 90 | |
BC006862.1 | - | 2 - 3 | 303 | 89 | |
BC013533.1 | - | 2 - 3 | 301 | 89 | |
CT010350.1 | - | 2 - 3 | 283 | 89 |