The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026175.5 | NP_080451.4 | 13 - 14 | 2231 | 139 |
GenBank mRNA | AK004763.1 | - | 13 - 14 | 2214 | 139 |
AK008075.1 | - | 2 - 3 | 258 | 139 | |
AK028829.1 | - | 13 - 14 | 2183 | 139 | |
AK029095.1 | - | 13 - 14 | 2201 | 139 | |
AK030115.1 | - | 5 - 6 | 757 | 139 | |
AK030223.1 | - | 13 - 14 | 2218 | 139 | |
AK134178.1 | - | 13 - 14 | 2209 | 139 | |
BC010727.1 | - | 4 - 5 | 686 | 139 | |
BC029753.1 | - | 13 - 14 | 2195 | 139 |