The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026225.3 | NP_080501.2 | 1 - 2 | 173 | 64 |
GenBank mRNA | AB093286.1 | - | 1 - 2 | 145 | 64 |
AK016663.1 | - | 1 - 2 | 186 | 64 | |
AK031149.1 | - | 1 - 2 | 167 | 64 | |
AK045223.1 | - | 1 - 2 | 167 | 64 | |
AK048986.1 | - | 1 - 2 | 160 | 64 | |
AK153320.1 | - | 1 - 2 | 162 | 64 | |
BC025427.1 | - | 1 - 2 | 179 | 64 |