Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kctd5
Entrez Gene ID
69259
Gene Name
potassium channel tetramerisation domain containing 5
Gene Aliases
2610030N08Rik, mKIAA0176
Chromosome Location
Chr.17: 24047720 - 24073485 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK129077.1 - 2 - 3 376 139
BC021777.1 - 2 - 3 368 139

View Full Product Details