The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027545.2 | NP_081821.1 | 16 - 17 | 2432 | 101 |
GenBank mRNA | AK033457.1 | - | 9 - 10 | 1176 | 101 |
AK045423.1 | - | 16 - 17 | 2433 | 101 | |
AK083386.1 | - | 16 - 17 | 2445 | 101 | |
AK167727.1 | - | 9 - 10 | 1155 | 101 | |
BC043480.1 | - | 16 - 17 | 2399 | 101 | |
BC125483.1 | - | 16 - 17 | 2469 | 101 | |
BC125485.1 | - | 16 - 17 | 2469 | 101 |