Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dzip1l
Entrez Gene ID
72507
Gene Name
DAZ interacting protein 1-like
Gene Aliases
2610524A10Rik, AI663976
Chromosome Location
Chr.9: 99629501 - 99669263 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028258.3 NP_082534.2 2 - 3 657 61
GenBank mRNA AK012132.1 - 3 - 4 1080 61
AK028787.1 - 3 - 4 812 61
AK034503.1 - 2 - 3 703 61
AK166083.1 - 3 - 4 815 61
BC099950.1 - 2 - 3 657 61
BC138218.1 - 1 - 2 586 61
BC145204.1 - 1 - 2 586 61

View Full Product Details