Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pih1d2
Entrez Gene ID
72614
Gene Name
PIH1 domain containing 2
Gene Aliases
2700059L22Rik
Chromosome Location
Chr.9: 50617321 - 50625000 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028300.2 NP_082576.1 4 - 5 849 66
GenBank mRNA AK012458.1 - 4 - 5 825 66
BC019481.1 - 4 - 5 747 66
BC039645.1 - 4 - 5 839 66

View Full Product Details