Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Usp20
Entrez Gene ID
74270
Gene Name
ubiquitin specific peptidase 20
Gene Aliases
1700055M05Rik, AI467231, Vdu2
Chromosome Location
Chr.2: 30982279 - 31022655 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028846.5 NP_083122.1 7 - 8 663 73
GenBank mRNA AF449715.1 - 7 - 8 614 73
AK029890.1 - - 560 73
AK042612.1 - 7 - 8 580 73
AK047242.1 - 7 - 8 582 73
AK054279.1 - 7 - 8 606 73
AK087412.1 - 7 - 8 574 73
AK163663.1 - 7 - 8 573 73
AK173083.1 - 7 - 8 565 73
BC037199.1 - 1 - 2 47 73
BC079674.1 - - 552 73

View Full Product Details