Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd55
Entrez Gene ID
77318
Gene Name
ankyrin repeat domain 55
Gene Aliases
C030011J08Rik
Chromosome Location
Chr.13: 112288451 - 112384002 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168403.1 NP_001161875.1 6 - 7 1058 66
NM_001168404.1 NP_001161876.1 6 - 7 595 66
NM_029898.3 NP_084174.2 5 - 6 952 66
GenBank mRNA AK045480.1 - 5 - 6 586 66
AK081197.1 - 6 - 7 596 66
AK138273.1 - 5 - 6 607 66
AK140417.1 - 6 - 7 922 66

View Full Product Details