Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nolc1
Entrez Gene ID
70769
Gene Name
nucleolar and coiled-body phosphoprotein 1
Gene Aliases
NOPP130, NOPP140, P130
Chromosome Location
Chr.19: 46075847 - 46085543 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039351.2 NP_001034440.1 4 - 5 635 87
NM_053086.3 NP_444316.2 4 - 5 638 87
GenBank mRNA AK014336.1 - 4 - 5 540 87
AK029150.1 - 4 - 5 486 87
AK034817.1 - 4 - 5 515 87
AK150332.1 - 4 - 5 472 87
AK151031.1 - 4 - 5 472 87
AK152941.1 - 4 - 5 472 87
AK166753.1 - 4 - 5 544 87
BC003244.1 - 4 - 5 481 87

View Full Product Details