The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271873.1 | NP_001258802.1 | 11 - 12 | 1755 | 85 |
NM_133979.3 | NP_598740.1 | 11 - 12 | 1929 | 85 | |
GenBank mRNA | AK028391.1 | - | 6 - 7 | 873 | 85 |
AK043011.1 | - | 11 - 12 | 1817 | 85 | |
AK046091.1 | - | 11 - 12 | 1881 | 85 | |
AK054163.1 | - | 7 - 8 | 1219 | 85 | |
AK080947.1 | - | 11 - 12 | 1881 | 85 | |
AK138414.1 | - | 11 - 12 | 2004 | 85 | |
AK186643.1 | - | 2 - 3 | 249 | 85 | |
BC002294.1 | - | 2 - 3 | 331 | 85 | |
BC026421.1 | - | 6 - 7 | 969 | 85 | |
BC099688.1 | - | 11 - 12 | 1678 | 85 |