The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164563.1 | NP_001158035.1 | 2 - 2 | 1923 | 87 |
NM_001164602.1 | NP_001158074.1 | 2 - 2 | 1891 | 87 | |
NM_178114.4 | NP_835215.1 | 1 - 1 | 2266 | 87 | |
GenBank mRNA | AB078880.1 | - | 1 - 1 | 1924 | 87 |
AK030848.1 | - | 1 - 1 | 2235 | 87 | |
AK038783.1 | - | 2 - 2 | 1925 | 87 | |
AY237006.1 | - | 1 - 1 | 1518 | 87 | |
BC095990.1 | - | 1 - 1 | 2101 | 87 |