Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brip1
Entrez Gene ID
237911
Gene Name
BRCA1 interacting protein C-terminal helicase 1
Gene Aliases
3110009N10Rik, 8030460J03Rik, Bach1, FACJ, Fancj, OF
Chromosome Location
Chr.11: 86058131 - 86201267 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178309.2 NP_840094.1 19 - 20 3023 106
GenBank mRNA AK033205.1 - 19 - 20 2994 106
AK041841.1 - 4 - 5 597 106
AK080771.1 - 4 - 5 650 106
BC094252.1 - 19 - 20 2972 106

View Full Product Details