Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Reep4
Entrez Gene ID
72549
Gene Name
receptor accessory protein 4
Gene Aliases
2700029E10Rik
Chromosome Location
Chr.14: 70545218 - 70548935 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180588.2 NP_850919.1 4 - 5 756 66
GenBank mRNA AK012301.1 - 4 - 5 655 66
AY562232.1 - 4 - 5 300 66
BC033929.1 - 4 - 5 384 66

View Full Product Details